missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine-4-carboxylic acid, 95%, Thermo Scientific™

£15.67 - £46.40

Chemical Identifiers

CAS 88912-26-9
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00466640
InChI Key GFOVTTQVBDEYPP-UHFFFAOYSA-N
Synonym 2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
PubChem CID 2736087
IUPAC Name 2,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)Cl)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484598
View Documents
Thermo Scientific Alfa Aesar
H58277.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15474598
View Documents
Thermo Scientific Alfa Aesar
H58277.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

88912-26-9
191.995
GFOVTTQVBDEYPP-UHFFFAOYSA-N
2736087
C1=C(C(=CN=C1Cl)Cl)C(=O)O
C6H3Cl2NO2
MFCD00466640
2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
2,5-dichloropyridine-4-carboxylic acid

Specifications

88912-26-9
C6H3Cl2NO2
2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
C1=C(C(=CN=C1Cl)Cl)C(=O)O
191.995
192
2,5-Dichloropyridine-4-carboxylic acid
235°C to 240°C
MFCD00466640
GFOVTTQVBDEYPP-UHFFFAOYSA-N
2,5-dichloropyridine-4-carboxylic acid
2736087
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications