missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine, 98%, Thermo Scientific Chemicals

Product Code. 10367220
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color White to Yellow
Density 112g/mL
Boiling Point 190.0°C to 191.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Cl2
Quantity 5 g
Beilstein 20, I, 80
Solubility Information Solubility in water: insoluble
Formula Weight 147.99
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2, 5-Dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'