missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-methylbenzoic acid, 95%, Thermo Scientific™

Product Code. 15440459
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103877-80-1
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD08063230
InChI Key PWWMQUUDAAWEBK-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
PubChem CID 13657451
IUPAC Name 2,5-difluoro-4-methylbenzoic acid
SMILES CC1=C(F)C=C(C(O)=O)C(F)=C1

Specifications

Color White
Quantity 250 mg
Formula Weight 172.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-4-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.