missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzophenone, 97+%, Thermo Scientific™

Product Code. 11449987
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-36-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00009899
InChI Key HSCUAAMDKDZZKG-UHFFFAOYSA-N
Synonym 2,5-difluorobenzophenone, 2,5-difluorophenyl phenyl methanone, 2,5-difluoro-phenyl-phenyl-methanone, methanone, 2,5-difluorophenyl phenyl, 2,5-difluoro diphenylmethanone, 2,5-difluorophenyl-phenylmethanone, 2,5-difluorophenyl phenyl ketone, pubchem4214, 2.5-difluorobenzophenone, 2,5-difluoro benzophenone
PubChem CID 522826
IUPAC Name (2,5-difluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)F)F

Specifications

Density 1.26
Boiling Point 120°C to 123°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5693
Quantity 5 g
Beilstein 4417185
Formula Weight 218.2
Percent Purity ≥97%
Chemical Name or Material 2,5-Difluorobenzophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'