missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dihydroxy-1,4-dithiane, 96%, Thermo Scientific Chemicals Product Code.: 11457566

2,5-Dihydroxy-1,4-dithiane, 96%, Thermo Scientific Chemicals

Product Code. 11457566
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11457566

Brand: Thermo Scientific Alfa Aesar A14620.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40018-26-6
Molecular Formula C4H8O2S2
Molecular Weight (g/mol) 152.226
MDL Number MFCD00006659
InChI Key YUIOPHXTILULQC-UHFFFAOYSA-N
Synonym p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anmeShow MoreShow Less
PubChem CID 98330
IUPAC Name 1,4-dithiane-2,5-diol
SMILES C1C(SCC(S1)O)O

Specifications

Melting Point ∼130°C (decomposition)
Quantity 50 g
UN Number UN2811
Beilstein 104402
Formula Weight 152.24
Percent Purity 96%
Chemical Name or Material 2,5-Dihydroxy-1,4-dithiane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251131 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14620.30,A14620.0B,A14620.18
Lot Number10219851 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberA14620.30,A14620.0B,A14620.18
Lot Number10223749 Certificate TypeCertificate of Analysis Date21/06/2022 Catalog NumberA14620.30,A14620.0B,A14620.18
Lot Number10122184 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14620.30,A14620.0B,A14620.18
Lot Number10202200 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14620.30,A14620.0B,A14620.18
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dihydroxy-1,4-dithiane, Practical Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Dithiane-2,5-diol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dihydroxy-1,4-dithiane, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.