Learn More
2,5-Dimethoxyaniline, 99%, Thermo Scientific Chemicals
£13.74 - £78.80
Chemical Identifiers
CAS | 102-56-7 |
---|---|
Molecular Formula | C8H11NO2 |
Molecular Weight (g/mol) | 153.18 |
MDL Number | MFCD00008368 |
InChI Key | NAZDVUBIEPVUKE-UHFFFAOYSA-N |
Synonym | benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline |
PubChem CID | 7613 |
IUPAC Name | 2,5-dimethoxyaniline |
SMILES | COC1=CC=C(OC)C(N)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10437772
|
Thermo Scientific Acros
150850050 |
5 g | Glass bottle | ||||||
10225910
|
Thermo Scientific Acros
150850500 |
50 g | Glass bottle | ||||||
10724081
|
Thermo Scientific Acros
150852500 |
250 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
102-56-7 | |
153.18 | |
NAZDVUBIEPVUKE-UHFFFAOYSA-N | |
7613 | |
COC1=CC=C(OC)C(N)=C1 |
C8H11NO2 | |
MFCD00008368 | |
benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline | |
2,5-dimethoxyaniline |
Specifications
102-56-7 | |
100.0 | |
Brown to Gray | |
130°C | |
98.5% min. (GC) | |
(CH3O)2C6H3NH2 | |
13, 788 | |
Solubility in water: 4g/L (40°C). Other solubilities: soluble in most common organic solvents | |
COC1=CC=C(OC)C(N)=C1 | |
153.18 | |
153.18 | |
Crystalline Powder or Lumps |
99.0 | |
79.0°C to 82.0°C | |
140.0°C (7.0 mmHg) | |
Authentic | |
C8H11NO2 | |
MFCD00008368 | |
benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline | |
NAZDVUBIEPVUKE-UHFFFAOYSA-N | |
2,5-dimethoxyaniline | |
7613 | |
99% | |
2, 5-Dimethoxyaniline, 99% |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-040-9
missing translation for 'rtecsNumber' : BX4375000
missing translation for 'tsca' : TSCA
RUO – Research Use Only