missing translation for 'onlineSavingsMsg'
Learn More

2,5-dimethoxyphenyl isothiocyanate, Thermo Scientific™

£18.70 - £62.10

Chemical Identifiers

CAS 40532-06-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00041059
InChI Key RADQLZMDTYWZKA-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
PubChem CID 142466
IUPAC Name 2-isothiocyanato-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(=C1)N=C=S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10352325
View Documents
Thermo Scientific Maybridge
SB01666DE
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10037592
View Documents
Thermo Scientific Maybridge
SB01666EA
10 g
£31.10
10g
Please to purchase this item. Need a web account? Register with us today!
10623275
View Documents
Thermo Scientific Maybridge
25 g
£62.10
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

40532-06-7
195.24
RADQLZMDTYWZKA-UHFFFAOYSA-N
142466
COC1=CC=C(OC)C(=C1)N=C=S
C9H9NO2S
MFCD00041059
2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
2-isothiocyanato-1,4-dimethoxybenzene

Specifications

40532-06-7
Amber glass bottle
MFCD00041059
RADQLZMDTYWZKA-UHFFFAOYSA-N
2-isothiocyanato-1,4-dimethoxybenzene
142466
97%
97%
C9H9NO2S
2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
COC1=CC=C(OC)C(=C1)N=C=S
195.24
195.24
2, 5-Dimethoxyphenyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications