missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

Product Code. 15401066
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62890-98-6
Molecular Formula C8H9BrMgO2
MDL Number MFCD01311475
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 2,5-dimethoxyphenylmagnesium bromide, 2,5-dimethoxyphenyl magnesium bromide, 2,5-dimethoxyphenylmagnesium bromide solution, grscplvzimbkky-uhfffaoysa-m, 2,5-dimethoxylphenylmagnesium bromide, magnesium,1,4-dimethoxybenzene-6-ide,bromide, 2,5-dimethoxyphenylmagnesium bromide, 0.5m in 2-methf, 2,5-dimethoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,5-dimethoxyphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
InChI Key GRSCPLVZIMBKKY-UHFFFAOYSA-M
SMILES COC1=CC=C(OC)C([Mg]Br)=C1
IUPAC Name magnesium(2+) 2,5-dimethoxybenzen-1-ide bromide
Molecular Weight (g/mol) 241.37
PubChem CID 11085924
Formula Weight 241.37
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 2,5-Dimethoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'