missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-1-(2-thienylmethyl)-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™
£23.00 - £77.15
Chemical Identifiers
CAS | 306936-14-1 |
---|---|
Molecular Formula | C12H13NO2S |
Molecular Weight (g/mol) | 235.301 |
MDL Number | MFCD01567217 |
InChI Key | MYRBXKXOHIDTFU-UHFFFAOYSA-N |
Synonym | 2,5-dimethyl-1-2-thienylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-2-thienylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thien-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-yl methyl-1h-pyrrole-3-carboxylic acid, maybridge1_008501, 2,5-dimethyl-1-thiophen-2-ylmethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-2-thienylmethyl, 2,5-dimethyl-1-2-thienyl-methyl-1h-pyrrole-3-carboxylic acid |
PubChem CID | 2736309 |
IUPAC Name | 2,5-dimethyl-1-(thiophen-2-ylmethyl)pyrrole-3-carboxylic acid |
SMILES | CC1=CC(=C(N1CC2=CC=CS2)C)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10509982
|
Thermo Scientific Maybridge
SP00291DA |
1 g |
£23.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10331925
|
Thermo Scientific Maybridge
SP00291DE |
5 g |
£77.15
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
306936-14-1 | |
235.301 | |
MYRBXKXOHIDTFU-UHFFFAOYSA-N | |
2736309 | |
CC1=CC(=C(N1CC2=CC=CS2)C)C(=O)O |
C12H13NO2S | |
MFCD01567217 | |
2,5-dimethyl-1-2-thienylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-2-thienylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thien-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-yl methyl-1h-pyrrole-3-carboxylic acid, maybridge1_008501, 2,5-dimethyl-1-thiophen-2-ylmethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-2-thienylmethyl, 2,5-dimethyl-1-2-thienyl-methyl-1h-pyrrole-3-carboxylic acid | |
2,5-dimethyl-1-(thiophen-2-ylmethyl)pyrrole-3-carboxylic acid |
Specifications
306936-14-1 | |
97% | |
C12H13NO2S | |
2,5-dimethyl-1-2-thienylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-2-thienylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thien-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-yl methyl-1h-pyrrole-3-carboxylic acid, maybridge1_008501, 2,5-dimethyl-1-thiophen-2-ylmethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-2-thienylmethyl, 2,5-dimethyl-1-2-thienyl-methyl-1h-pyrrole-3-carboxylic acid | |
CC1=CC(=C(N1CC2=CC=CS2)C)C(=O)O | |
235.301 | |
235.31 | |
2, 5-Dimethyl-1-(2-thienylmethyl)-1H-pyrrole-3-carboxylic acid |
97.0 | |
Amber Glass Bottle | |
MFCD01567217 | |
MYRBXKXOHIDTFU-UHFFFAOYSA-N | |
2,5-dimethyl-1-(thiophen-2-ylmethyl)pyrrole-3-carboxylic acid | |
2736309 | |
97% |