missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10572852
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 197719-27-0
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.57
MDL Number MFCD04972642
InChI Key XZXCVPKMCPXVFF-UHFFFAOYSA-N
PubChem CID 7018020
SMILES CC1=NC(C(Cl)=O)=C(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
IUPAC Name 2,5-dimethyl-1,3-oxazole-4-carbonyl chloride
Formula Weight 159.57
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'