missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals

Product Code. 10153191 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 10153191

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-63-7
Molecular Formula C16H34O4
Molecular Weight (g/mol) 290.44
InChI Key DMWVYCCGCQPJEA-UHFFFAOYSA-N
Synonym varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
PubChem CID 6545
IUPAC Name 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
SMILES CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C

Specifications

Melting Point 6.0°C
Density 0.8700g/mL
Boiling Point 55.0°C to 57.0°C (7.0 mmHg)
Flash Point 68°C
Assay Percent Range 90% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4200 to 1.4240
Quantity 100 g
Specific Gravity 0.87
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in organic solvents,soluble in alcohol
Viscosity 6.4 mPa.s (20°C)
Formula Weight 290.44
Percent Purity 92%
Chemical Name or Material 2, 5-Dimethyl-2, 5-di(tert-butylperoxy)hexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane
Signal Word
  • Danger
Hazard Category
  • Organic peroxide Type C & D
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.