missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-hexanol, 98%, Thermo Scientific™

Product Code. 11458967
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 19550-07-3
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00026452
InChI Key SNKTZHPOKPYBPT-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-hexanol, 3-hexanol, 2,5-dimethyl, acmc-209ezm, 3-hexanol,2,5-dimethyl
PubChem CID 89183
IUPAC Name 2,5-dimethylhexan-3-ol
SMILES CC(C)CC(C(C)C)O

Specifications

Density 0.82
Quantity 1 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 2,5-Dimethyl-3-hexanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethyl-3-hexanol, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.