missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylbenzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | (CH3)2C6H3SO2Cl |
Formula Weight | 204.68 |
Percent Purity | 97% |
Quantity | 1 g |
Packaging | Amber Glass Bottle |
Chemical Name or Material | 2, 5-Dimethylbenzenesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'