Learn More
2,5-Dimethylbenzenesulfonyl chloride, 98%, Thermo Scientific™
£24.30 - £50.90
Chemical Identifiers
CAS | 19040-62-1 |
---|---|
Molecular Formula | C8H9ClO2S |
Molecular Weight (g/mol) | 204.668 |
MDL Number | MFCD00024875 |
InChI Key | FZVZUIBYLZZOEW-UHFFFAOYSA-N |
Synonym | p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f |
PubChem CID | 87910 |
IUPAC Name | 2,5-dimethylbenzenesulfonyl chloride |
SMILES | CC1=CC(=C(C=C1)C)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316229
|
Thermo Scientific Alfa Aesar
044826.03 |
1 g |
£24.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326229
|
Thermo Scientific Alfa Aesar
044826.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19040-62-1 | |
204.668 | |
FZVZUIBYLZZOEW-UHFFFAOYSA-N | |
87910 | |
CC1=CC(=C(C=C1)C)S(=O)(=O)Cl |
C8H9ClO2S | |
MFCD00024875 | |
p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f | |
2,5-dimethylbenzenesulfonyl chloride |
Specifications
20°C | |
Yellow | |
Liquid/Solid | |
MFCD00024875 | |
p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f | |
CC1=CC(=C(C=C1)C)S(=O)(=O)Cl | |
204.668 | |
204.68 | |
2,5-Dimethylbenzenesulfonyl chloride |
19040-62-1 | |
120°C to 125°C | |
C8H9ClO2S | |
UN3265 | |
FZVZUIBYLZZOEW-UHFFFAOYSA-N | |
2,5-dimethylbenzenesulfonyl chloride | |
87910 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : DB8927600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen