missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one, 97%, Thermo Scientific™

Catalog Number 11419926
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
98488-10-9
C8H9N3O
163.18
MFCD08729601
LGQMDCIHBYTQJU-UHFFFAOYSA-N
2,5-dimethylpyrazolo 1,5-a pyrimidin-7-ol, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7 4h-one, 2,5-dimethyl-4h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4,7-dihydropyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-1h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4h,7h-pyrazolo 1,5-a pyrimidin-7-one, acmc-20aoa7, maybridge1_004141
736199
2,5-dimethyl-1H-pyrazolo[1,5-a]pyrimidin-7-one
CC1=CC2=NC(C)=CC(=O)N2N1
This item is not returnable. View return policy
98488-10-9
C8H9N3O
163.18
MFCD08729601
LGQMDCIHBYTQJU-UHFFFAOYSA-N
2,5-dimethylpyrazolo 1,5-a pyrimidin-7-ol, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7 4h-one, 2,5-dimethyl-4h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4,7-dihydropyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-1h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4h,7h-pyrazolo 1,5-a pyrimidin-7-one, acmc-20aoa7, maybridge1_004141
736199
2,5-dimethyl-1H-pyrazolo[1,5-a]pyrimidin-7-one
CC1=CC2=NC(C)=CC(=O)N2N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 252°C to 254°C
Quantity 1 g
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'