missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals

Product Code. 11483394
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym 2,5-diphenyloxazole, oxazole, 2,5-diphenyl, tritosol, 2,5-diphenyl-oxazole, ppo scintillator, dpo scintillator, usaf ek-6775, ppo van, ppo, ppo scintillator van
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C to 74°C
Density 1.06
Boiling Point 360°C
Quantity 25 g
Beilstein 157021
Formula Weight 221.26
Percent Purity 99%
Chemical Name or Material 2,5-Diphenyloxazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.