Learn More
Fisher Chemical 2.5 LT ETHANOL ABSOLUTE PRIMAR 2.5LITRE
Brand: Fisher Chemical E/0655/17
2.5 LT ETHANOL ABSOLUTE PRIMAR 2.5LITRE
This item is not returnable.
View return policy
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol, absolute.
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Product Title
Fisher Chemical 2.5 LT ETHANOL ABSOLUTE PRIMAR 2.5LITRE >
Spot an opportunity for improvement?Share a Content Correction