missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Chemical 2.5 LT ETHANOL AR 95% IN PLASTIC CONTAINER 2.5L
Brand: Fisher Chemical E/0550/PC17
2.5 LT ETHANOL AR 95% IN PLASTIC CONTAINER 2.5L
This item is not returnable.
View return policy
Description
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol 95% v/v.
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher Chemical 2.5 LT ETHANOL AR 95% IN PLASTIC CONTAINER 2.5L >
Spot an opportunity for improvement?Share a Content Correction