missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Quantity | 25 g |
| Formula Weight | 217.22 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)acetic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction