Learn More
2.5LT Ethanol absolute, 99.8+%, for analysis
Brand: Fisher Chemical E/0650/PC17
2.5LT Ethanol absolute, 99.8+%, for analysis
Code : 22
Additional Details : CAS Number : 64-17-5 Weight : 1.97250kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Safety and Handling
- Ethanol, absolute
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.