Learn More
2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
£26.60 - £84.50
Chemical Identifiers
CAS | 24821-22-5 |
---|---|
Molecular Formula | C9H4F6O2 |
Molecular Weight (g/mol) | 258.11 |
InChI Key | XZNLSDPNMNWCRE-UHFFFAOYSA-N |
Synonym | 2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962 |
PubChem CID | 90618 |
IUPAC Name | 2,6-bis(trifluoromethyl)benzoic acid |
SMILES | C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10189629
|
Thermo Scientific Acros
214460010 |
1 g | Glass bottle |
£26.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10129399
|
Thermo Scientific Acros
214460050 |
5 g | Glass Bottle |
£84.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24821-22-5 | |
258.11 | |
2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962 | |
2,6-bis(trifluoromethyl)benzoic acid |
C9H4F6O2 | |
XZNLSDPNMNWCRE-UHFFFAOYSA-N | |
90618 | |
C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F |
Specifications
24821-22-5 | |
100.0 | |
White | |
97.5% min. (HPLC) | |
(CF3)2C6H3CO2H | |
XZNLSDPNMNWCRE-UHFFFAOYSA-N | |
2,6-bis(trifluoromethyl)benzoic acid | |
90618 | |
98% | |
2, 6-Bis(trifluoromethyl)benzoic acid |
97.5 | |
137.7°C to 138.6°C | |
Authentic | |
C9H4F6O2 | |
2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962 | |
C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F | |
258.11 | |
258.11 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-479-1
RUO – Research Use Only