missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11466276
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53130-44-2
Molecular Formula C9H3ClF6O
Molecular Weight (g/mol) 276.562
MDL Number MFCD00061228
InChI Key NQMCJGTVZMBTDS-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl benzoyl chloride, pubchem10021, acmc-1aoxm, 2,6-bis-trifluoromethylbenzoyl chloride, 2,6-bis-trifluoromethyl-benzoyl chloride, benzoylchloride, 2,6-bis trifluoromethyl, benzoyl chloride, 2,6-bis trifluoromethyl, 2,6-bis trifluoromethyl benzoylchloride
PubChem CID 2736128
IUPAC Name 2,6-bis(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=C(C(=C1)C(F)(F)F)C(=O)Cl)C(F)(F)F

Specifications

Refractive Index 1.4304
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 276.57
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'