missing translation for 'onlineSavingsMsg'
Learn More
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals Product Code.: 10339250

2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals

Product Code. 10339250
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10339250

Brand: Thermo Scientific Acros 294161000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489-01-0
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.35
MDL Number MFCD00008824
InChI Key SLUKQUGVTITNSY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253
PubChem CID 10269
IUPAC Name 2,6-ditert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 103°C to 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 100 g
Fieser 16,110; 16,58
Solubility Information Solubility in water: practically insoluble
Formula Weight 236.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Di-tert-butyl-4-methoxyphenol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Di-tert-butyl-4-methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.