Learn More
2,6-Di-tert-butylpyridine, 97+%, Thermo Scientific Chemicals
£32.70 - £372.00
Chemical Identifiers
CAS | 585-48-8 |
---|---|
Molecular Formula | C13H21N |
Molecular Weight (g/mol) | 191.318 |
MDL Number | MFCD00006306 |
InChI Key | UWKQJZCTQGMHKD-UHFFFAOYSA-N |
Synonym | 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 |
PubChem CID | 68510 |
IUPAC Name | 2,6-ditert-butylpyridine |
SMILES | CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492098
|
Thermo Scientific Alfa Aesar
H55761.03 |
1 g |
£32.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402108
|
Thermo Scientific Alfa Aesar
H55761.06 |
5 g |
£108.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412108
|
Thermo Scientific Alfa Aesar
H55761.14 |
25 g |
£372.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.
Solubility
Miscible with alcohol, acetone, and hexane. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
Chemical Identifiers
585-48-8 | |
191.318 | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
68510 | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
C13H21N | |
MFCD00006306 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
2,6-ditert-butylpyridine |
Specifications
585-48-8 | |
100°C to 101°C (23 mmHg) | |
C13H21N | |
MFCD00006306 | |
14,3036 | |
Miscible with alcohol,acetone,and hexane. Immiscible with water. | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C | |
191.318 | |
191.32 | |
2,6-Di-tert-butylpyridine |
0.867 | |
71°C (160°F) | |
1.473 | |
125886 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
2,6-ditert-butylpyridine | |
68510 | |
≥97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 209-557-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only