missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminopimelic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.
Solubility
Soluble in water, diluted acids and alkali.
Notes
Incompatible with strong oxidizing agents.
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.
Solubility
Soluble in water, diluted acids and alkali.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C |
Assay Percent Range | 0.98 |
Quantity | 1 g |
Solubility Information | Soluble in water,diluted acids and alkali. |
Formula Weight | 190.2 |
Chemical Name or Material | 2, 6-Diaminopimelic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'