missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopimelic acid, 96%, Thermo Scientific Chemicals

Product Code. 11460507
Click to view available options
Quantity:
1 g
5 g

2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

Solubility
Soluble in water, diluted acids and alkali.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-93-7
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.199
MDL Number MFCD00002637
InChI Key GMKMEZVLHJARHF-UHFFFAOYSA-N
Synonym 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
PubChem CID 865
ChEBI CHEBI:23673
IUPAC Name 2,6-diaminoheptanedioic acid
SMILES C(CC(C(=O)O)N)CC(C(=O)O)N

Specifications

Melting Point >300°C
Assay Percent Range 0.98
Quantity 5 g
Solubility Information Soluble in water,diluted acids and alkali.
Formula Weight 190.2
Chemical Name or Material 2, 6-Diaminopimelic Acid
compliance-icons
Product Identifier
  • 2,6-Diaminopimelic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Diaminopimelic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'