missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-methylaniline, 98+%

Product Code. 11433407
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11433407

Brand: Thermo Scientific Alfa Aesar A17184.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6968-24-7 | C7H7Br2N | 264.948 g/mol

2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

Solubility
Insoluble in water.

Notes
Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6968-24-7
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00007641
InChI Key ATDIROHVRVQMRO-UHFFFAOYSA-N
Synonym 2,6-dibromo-p-toluidine, 3,5-dibromo-4-aminotoluene, benzenamine, 2,6-dibromo-4-methyl, 2,6-dibromo-4-methyl aniline, 2,6-dibromo-4-methyl-phenylamine, 2,6-dibromo-4-methylphenylamine, 2,6-dibromo-4-methyl-benzenamine, pubchem2334, acmc-209oag, 2,6-dibromo-p-toludine
PubChem CID 81427
IUPAC Name 2,6-dibromo-4-methylaniline
SMILES CC1=CC(=C(C(=C1)Br)N)Br

Specifications

Melting Point 72°C to 76°C
Quantity 25 g
Beilstein 2085782
Solubility Information Insoluble in water.
Formula Weight 264.96
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromo-4-methylaniline
compliance-icons
Product Identifier
  • 2,6-Dibromo-4-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Dibromo-4-methylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.