missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromopyrazine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Color | White to Brown |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 237.88 |
Percent Purity | 95% |
Physical Form | Crystalline Solid |
Chemical Name or Material | 2,6-Dibromopyrazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'