missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-(chloromethyl)pyridine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 196.46 |
Percent Purity | 95% |
Chemical Name or Material | 2, 6-Dichloro-4-(chloromethyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'