missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dichloro-4-nitroaniline, 95%, Thermo Scientific Chemicals Product Code.: 11494060

2,6-Dichloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Product Code. 11494060
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11494060

Brand: Thermo Scientific Alfa Aesar B24283.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as dyestuff, agricultural and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007677
InChI Key BIXZHMJUSMUDOQ-UHFFFAOYSA-N
Synonym dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide
PubChem CID 7430
ChEBI CHEBI:27864
IUPAC Name 2,6-dichloro-4-nitroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]

Specifications

Melting Point 185°C to 192°C
Boiling Point 130°C (2 mmHg)
Odor Recognizable
Quantity 25 g
Beilstein 1459581
Solubility Information Insoluble in water.
Formula Weight 207.02
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-nitroaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU23K036 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberB24283.22,B24283.14,B24283.36
Lot Number10241201 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog NumberB24283.22,B24283.14,B24283.36
Lot Number.H9291A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24283.22,B24283.14,B24283.36
Lot NumberW29C049 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24283.22,B24283.14,B24283.36
Lot NumberP10G111 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24283.22,B24283.14,B24283.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dichloro-4-nitroaniline, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.