missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals

£40.30 - £145.00

Chemical Identifiers

CAS 2040-05-3
MDL Number MFCD00016339
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302486
View Documents
Thermo Scientific Alfa Aesar
L06348.03
1 g
£40.30
1g
Please to purchase this item. Need a web account? Register with us today!
11312486
View Documents
Thermo Scientific Alfa Aesar
L06348.06
5 g
£145.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2040-05-3
MFCD00016339

Specifications

2040-05-3
176°C to 180°C (83 mmHg)
MFCD00016339
2',4',5'-trimethylacetophenone, 1-2,4,5-trimethylphenyl ethanone, 1-2,4,5-trimethylphenyl ethan-1-one, acetophenone, 2',4',5'-trimethyl, ethanone, 1-2,4,5-trimethylphenyl, 1-acetyl-2,4,5-trimethylbenzene, acmc-20aofe, 2',5'-trimethylacetophenone, 2,4,5-trimethyl-acetophenone, acetophenone,4',5'-trimethyl
GENBEGZNCBFHSU-UHFFFAOYSA-N
1-(2,4,5-trimethylphenyl)ethanone
74878
98%
41°C to 44°C
C11H14O
2440104
Insoluble in water.
CC1=CC(=C(C=C1C(=O)C)C)C
162.23
189.04
2',6'-Dichloroacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

missing translation for 'einecsNumber' : 218-035-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only