Learn More
2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals
£20.70 - £136.00
Chemical Identifiers
CAS | 608-31-1 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.01 |
MDL Number | MFCD00007675 |
InChI Key | JDMFXJULNGEPOI-UHFFFAOYSA-N |
Synonym | benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline |
PubChem CID | 11846 |
ChEBI | CHEBI:46630 |
IUPAC Name | 2,6-dichloroaniline |
SMILES | NC1=C(Cl)C=CC=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499423
|
Thermo Scientific Alfa Aesar
A12320.14 |
25 g |
£20.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409433
|
Thermo Scientific Alfa Aesar
A12320.22 |
100 g |
£41.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419433
|
Thermo Scientific Alfa Aesar
A12320.36 |
500 g |
£136.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.
Solubility
Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
608-31-1 | |
162.01 | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
11846 | |
2,6-dichloroaniline |
C6H5Cl2N | |
MFCD00007675 | |
benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline | |
CHEBI:46630 | |
NC1=C(Cl)C=CC=C1Cl |
Specifications
608-31-1 | |
97°C (5 mmHg) | |
C6H5Cl2N | |
UN3442 | |
benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline | |
NC1=C(Cl)C=CC=C1Cl | |
162.01 | |
CHEBI:46630 | |
98% |
37°C to 40°C | |
118°C (244°F) | |
MFCD00007675 | |
775084 | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
2,6-dichloroaniline | |
11846 | |
162.02 | |
2,6-Dichloroaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
missing translation for 'rtecsNumber' : CX9862655
missing translation for 'tsca' : TSCA
RUO – Research Use Only