missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzonitrile, 98%

Product Code. 11417207
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11417207

Brand: Thermo Scientific Alfa Aesar A14986.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1194-65-6 | C7H3Cl2N | 172.008 g/mol

2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

Solubility
Soluble in water (0.018g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-65-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00001781
InChI Key YOYAIZYFCNQIRF-UHFFFAOYSA-N
Synonym dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133
PubChem CID 3031
ChEBI CHEBI:943
IUPAC Name 2,6-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)Cl

Specifications

Melting Point 143°C to 147°C
Boiling Point 270°C
Odor Aromatic
Quantity 25 g
UN Number UN3077
Beilstein 1909167
Merck Index 14,3042
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzonitrile
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dichlorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H312-Harmful in contact with skin.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : Dichlobenil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.