missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichlorobenzyl)triphenylphosphonium chloride, 98%, Thermo Scientific™

Product Code. 11473218
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18583-54-5
Molecular Formula C25H20Cl3P
Molecular Weight (g/mol) 457.76
MDL Number MFCD00061392
InChI Key NBWPWZJEEQWREI-UHFFFAOYSA-M
Synonym 2,6-dichlorobenzyl triphenylphosphonium chloride, 2,6-dichlorophenyl methyl triphenylphosphanium chloride, acmc-20ao99, 2,6-dichlorobenzyltriphenylphosphoniumchloride, 2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride, 2,6-dichlorophenyl methyl triphenyl phosphanium chloride, phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1
PubChem CID 57370833
IUPAC Name (2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride
SMILES [Cl-].ClC1=CC=CC(Cl)=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 457.78
Percent Purity 98%
Chemical Name or Material (2,6-Dichlorobenzyl)triphenylphosphonium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'