missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorophenolindophenol Sodium Salt Hydrate, Honeywell Fluka™
Redox indicator
£40.59 - £219.32
Chemical Identifiers
CAS | 620-45-1 |
---|---|
Molecular Formula | C12H6Cl2NNaO2 |
Molecular Weight (g/mol) | 290.07 |
MDL Number | MFCD00150014 |
InChI Key | FHLDWQLHDYCXKI-UHFFFAOYSA-M |
PubChem CID | 23697355 |
ChEBI | CHEBI:948 |
SMILES | [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
620-45-1 | |
290.07 | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
CHEBI:948 |
C12H6Cl2NNaO2 | |
MFCD00150014 | |
23697355 | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Specifications
>300°C | |
C12H6Cl2NNaO2 | |
NONH for all modes of transport | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate | |
23697355 | |
290.08g/mol (anhydrous basis) | |
Odourless | |
Powder |
620-45-1 | |
MFCD00150014 | |
3641229 | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 | |
290.07 | |
CHEBI:948 | |
≤14% (120°C) | |
Green | |
2,6-Dichlorophenolindophenol Sodium Salt Hydrate |
Safety and Handling
missing translation for 'einecsNumber' : 210-640-4
missing translation for 'rtecsNumber' : GU5495000