missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenyl isothiocyanate, 98+%, Thermo Scientific Chemicals

Product Code. 11323306
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-95-0
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041056
InChI Key SUCGVQHNGIQXGD-UHFFFAOYSA-N
Synonym 2,6-dichlorophenyl isothiocyanate, 2,6-dichlorophenylisothiocyanate, benzene, 1,3-dichloro-2-isothiocyanato, 1,3-dichloro-2-isothiocyanato-benzene, benzene,1,3-dichloro-2-isothiocyanato, c7h3c12ns, 2,6-dichlorobenzenisothiocyanate, 2,6-dichlorophenyl-isothiocyanate, 1,3-dichloro-2-thioisocyanato-benzene, 2,6-dichloro-1-isothiocyanato-benzene
PubChem CID 123239
IUPAC Name 1,3-dichloro-2-isothiocyanatobenzene
SMILES ClC1=CC=CC(Cl)=C1N=C=S

Specifications

Melting Point 42°C to 44°C
Boiling Point 122°C to 124°C (8 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 639061
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity ≥98%
Chemical Name or Material 2,6-Dichlorophenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'