missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylboronic acid, 98%, Thermo Scientific™

Product Code. 10383032
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 73852-17-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 153°C to 154°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H5BCl2O2
MDL Number MFCD00064869
Quantity 500 mg
Synonym 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
InChI Key CXDPUSMFYPQXCV-UHFFFAOYSA-N
SMILES OB(O)C1=C(Cl)C=CC=C1Cl
IUPAC Name (2,6-dichlorophenyl)boronic acid
Molecular Weight (g/mol) 190.81
PubChem CID 2734332
Formula Weight 190.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 6-Dichlorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichlorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'