Learn More
2,6-Dichloropyrazine, 99+%, Thermo Scientific Chemicals
£24.40 - £256.00
Chemical Identifiers
CAS | 4774-14-5 |
---|---|
Molecular Formula | C4H2Cl2N2 |
Molecular Weight (g/mol) | 148.98 |
InChI Key | LSEAAPGIZCDEEH-UHFFFAOYSA-N |
Synonym | pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine |
PubChem CID | 78504 |
IUPAC Name | 2,6-dichloropyrazine |
SMILES | C1=C(N=C(C=N1)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772081
|
Thermo Scientific Acros
232540010 |
1 g | Glass bottle |
£24.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348320
|
Thermo Scientific Acros
232540050 |
5 g | Glass bottle |
£79.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056193
|
Thermo Scientific Acros
232540250 |
25 g | Glass bottle |
£256.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4774-14-5 | |
148.98 | |
pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine | |
2,6-dichloropyrazine |
C4H2Cl2N2 | |
LSEAAPGIZCDEEH-UHFFFAOYSA-N | |
78504 | |
C1=C(N=C(C=N1)Cl)Cl |
Specifications
4774-14-5 | |
100.0 | |
White to Beige | |
Authentic | |
C4H2Cl2N2 | |
pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine | |
C1=C(N=C(C=N1)Cl)Cl | |
148.98 | |
148.98 | |
Crystalline Powder or Crystals |
99.0 | |
53.0°C to 56.0°C | |
>110°C | |
99% min. (GC) | |
23, III, 903 | |
LSEAAPGIZCDEEH-UHFFFAOYSA-N | |
2,6-dichloropyrazine | |
78504 | |
99+% | |
2, 6-Dichloropyrazine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 225-316-8
RUO – Research Use Only