missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine, 98%, Thermo Scientific Chemicals

Product Code. 11410173
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2402-78-0
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006244
InChI Key FILKGCRCWDMBKA-UHFFFAOYSA-N
Synonym pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg
PubChem CID 16989
IUPAC Name 2,6-dichloropyridine
SMILES ClC1=CC=CC(Cl)=N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 211°C
Odor Unpleasant
Quantity 100 g
UN Number UN2811
Beilstein 108664
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'