missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorotoluene, 99%, Thermo Scientific Chemicals

£23.60 - £150.75

Chemical Identifiers

CAS 118-69-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000576
InChI Key DMEDNTFWIHCBRK-UHFFFAOYSA-N
Synonym 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
PubChem CID 8368
IUPAC Name 1,3-dichloro-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415927
View Documents
Thermo Scientific Alfa Aesar
B24463.14
25 g
£23.60
25g
Please to purchase this item. Need a web account? Register with us today!
11425927
View Documents
Thermo Scientific Alfa Aesar
B24463.22
100 g
£51.90
100g
Please to purchase this item. Need a web account? Register with us today!
11435927
View Documents
Thermo Scientific Alfa Aesar
B24463.36
500 g
£150.75
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

118-69-4
161.025
DMEDNTFWIHCBRK-UHFFFAOYSA-N
8368
CC1=C(C=CC=C1Cl)Cl
C7H6Cl2
MFCD00000576
2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
1,3-dichloro-2-methylbenzene

Specifications

118-69-4
1.254
82°C (179°F)
1.551
UN3082
2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
DMEDNTFWIHCBRK-UHFFFAOYSA-N
1,3-dichloro-2-methylbenzene
8368
99%
2°C
196°C to 203°C
C7H6Cl2
MFCD00000576
1907194
Soluble in water.
CC1=C(C=CC=C1Cl)Cl
161.025
161.03
2,6-Dichlorotoluene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 204-269-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only