Learn More
2,6-Dichlorotoluene, 99+%, Thermo Scientific Chemicals
£51.60 - £816.00
Chemical Identifiers
CAS | 118-69-4 |
---|---|
Molecular Formula | C7H6Cl2 |
Molecular Weight (g/mol) | 161.03 |
MDL Number | MFCD00000576 |
InChI Key | DMEDNTFWIHCBRK-UHFFFAOYSA-N |
Synonym | 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene |
PubChem CID | 8368 |
IUPAC Name | 1,3-dichloro-2-methylbenzene |
SMILES | CC1=C(C=CC=C1Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10590371
|
Thermo Scientific Acros
113791000 |
100 mL | Glass bottle |
£51.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10142380
|
Thermo Scientific Acros
113795000 |
500 mL | Glass bottle |
£168.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541184
|
Thermo Scientific Acros
113790025 |
2.5 L | Glass bottle |
£816.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
118-69-4 | |
161.03 | |
DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
8368 | |
CC1=C(C=CC=C1Cl)Cl |
C7H6Cl2 | |
MFCD00000576 | |
2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene | |
1,3-dichloro-2-methylbenzene |
Specifications
118-69-4 | |
100.0 | |
Colorless to Yellow | |
196°C to 203°C | |
Authentic | |
C7H6Cl2 | |
1.5500 to 1.552 | |
05, 296 | |
2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene | |
DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
1,3-dichloro-2-methylbenzene | |
8368 | |
99+% | |
2, 6-Dichlorotoluene, 99+% |
99.0 | |
2°C | |
1.2500g/mL | |
82°C | |
99% min. (GC) | |
CH3C6H3Cl2 | |
MFCD00000576 | |
1.25 | |
Solubility in water: 24mg/l (20°C). Other solubilities: soluble in alcohol,ether and acetone | |
CC1=C(C=CC=C1Cl)Cl | |
161.03 | |
161.03 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-269-7
missing translation for 'tsca' : TSCA
- 2,6-Dichlorotoluene
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only