missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diethylaniline, 98%, Thermo Scientific Chemicals

Product Code. 10317680
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-66-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00007753
InChI Key FOYHNROGBXVLLX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine
PubChem CID 11369
ChEBI CHEBI:27880
IUPAC Name 2,6-diethylaniline
SMILES CCC1=C(C(=CC=C1)CC)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 3°C to 4°C
Color Brown-Red to Yellow
Density 0.9000g/mL
Boiling Point 243°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2C6H3NH2
Refractive Index 1.5446 to 1.5466
Quantity 1 L
Beilstein 12,164
Specific Gravity 0.9
Formula Weight 149.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Diethylaniline, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'