missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methylphenylacetic acid, 97%, Thermo Scientific™

Product Code. 11491310
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261943-97-9
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD01631338
InChI Key XMNMLOQKAUVILT-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-methylphenylacetic acid, 2-2,6-difluoro-3-methylphenyl acetic acid, 2,6-difluoro-3-methylphenyl acetic acid, acmc-1ce69, benzeneacetic acid,2,6-difluoro-3-methyl
PubChem CID 2774155
IUPAC Name 2-(2,6-difluoro-3-methylphenyl)acetic acid
SMILES CC1=C(C(=C(C=C1)F)CC(=O)O)F

Specifications

Quantity 1 g
Formula Weight 186.16
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-3-methylphenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'