missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',6'-Difluoroacetophenone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2?,6?-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications
Specifications
Density | 1.195 |
Boiling Point | 76°C to 79°C (15 mmHg) |
Flash Point | 76°C (168°F) |
Refractive Index | 1.481 |
Quantity | 1 g |
Beilstein | 2046068 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 1-(2,6-difluorophenyl)ethan-1-one |
Formula Weight | 156.13 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'