missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzenesulfonamide, 97%, Thermo Scientific Chemicals

Product Code. 11476230
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60230-37-7
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.17
MDL Number MFCD00729101
InChI Key RVVVGGCOFWWDEL-UHFFFAOYSA-N
Synonym 2,6-difluorobenzene-1-sulfonamide, 2,6-difluorobenzenesulphonamide, benzenesulfonamide, 2,6-difluoro, fbt, 2,6-difluorophenylsulphonylamide, pubchem11779, acmc-209vki, d05dfj, ksc495s1r, 2,6-difluoro-benzenesulfonamide
PubChem CID 446274
ChEBI CHEBI:42429
IUPAC Name 2,6-difluorobenzenesulfonamide
SMILES NS(=O)(=O)C1=C(F)C=CC=C1F

Specifications

Melting Point 188°C to 192°C
Quantity 5 g
Formula Weight 193.17
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzenesulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'