missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11353056
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25343-70-8
Molecular Formula C13H17NS
Molecular Weight (g/mol) 219.346
MDL Number MFCD00041341
InChI Key HZGOUCYIYIFQHX-UHFFFAOYSA-N
Synonym 2,6-diisopropylphenyl isothiocyanate, 1,3-diisopropyl-2-isothiocyanatobenzene, 2-isothiocyanato-1,3-bis propan-2-yl benzene, 2,6-diisopropylphenylisothiocyanate, 1,3-diisopropyl-2-isothiocyanato-benzene, 2-isothiocyanato-1,3-di propan-2-yl benzene, acmc-1cue0, 2,6-diisopropylphenyl-isothiocyanate, 2,6-diisopropyl-phenyl isothiocyanate, 2,6-bis methylethyl benzenisothiocyanate
PubChem CID 141194
IUPAC Name 2-isothiocyanato-1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N=C=S

Specifications

Boiling Point 139°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 1 g
UN Number UN2922
Beilstein 2416576
Sensitivity Moisture sensitive
Formula Weight 219.35
Percent Purity 97%
Chemical Name or Material 2,6-Diisopropylphenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'