missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

£41.30 - £448.00

Chemical Identifiers

CAS 2040-04-2
MDL Number MFCD00008729
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11452738
View Documents
Thermo Scientific Alfa Aesar
A16733.06
5 g
£41.30
5g
Please to purchase this item. Need a web account? Register with us today!
11462738
View Documents
Thermo Scientific Alfa Aesar
25 g
£138.00
25g
Please to purchase this item. Need a web account? Register with us today!
11472738
View Documents
Thermo Scientific Alfa Aesar
A16733.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2040-04-2
MFCD00008729

Specifications

2040-04-2
135°C to 136°C (2 mmHg)
MFCD00008729
2',6'-dichloroacetophenone, 1-2,6-dichlorophenyl ethanone, 2,6-dichloroacetophenone, 1-2,6-dichlorophenyl ethan-1-one, ethanone, 1-2,6-dichlorophenyl, 2,6-dichloro acetophenone, 1-acetyl-2,6-dichlorobenzene, zlchem 1084, pubchem12362, acmc-1cobf
CC(=O)C1=C(C=CC=C1Cl)Cl
189.04
180.2
2',6'-Dimethoxyacetophenone
68°C to 71°C
C8H6Cl2O
2048976
HYBDSXBLGCQKRE-UHFFFAOYSA-N
1-(2,6-dichlorophenyl)ethanone
74877
98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 218-034-1

missing translation for 'rtecsNumber' : AM8400000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only