Learn More
2,6-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
£30.80 - £98.90
Chemical Identifiers
CAS | 1466-76-8 |
---|---|
Molecular Formula | C9H10O4 |
Molecular Weight (g/mol) | 182.18 |
MDL Number | MFCD00002437 |
InChI Key | MBIZFBDREVRUHY-UHFFFAOYSA-N |
Synonym | benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 |
PubChem CID | 15109 |
IUPAC Name | 2,6-dimethoxybenzoic acid |
SMILES | COC1=CC=CC(OC)=C1C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10162760
|
Thermo Scientific Acros
115440250 |
25 g | Glass bottle |
£30.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10774411
|
Thermo Scientific Acros
115441000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1466-76-8 | |
182.18 | |
MBIZFBDREVRUHY-UHFFFAOYSA-N | |
15109 | |
COC1=CC=CC(OC)=C1C(O)=O |
C9H10O4 | |
MFCD00002437 | |
benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
2,6-dimethoxybenzoic acid |
Specifications
1466-76-8 | |
100.0 | |
White | |
98.5% min. (HPLC) | |
(CH3O)2C6H3CO2H | |
10, 388 | |
Solubility in water: 0,35 wt% (25 c). Other solubilities: 0.29 wt% in benzene (25 c),3.64 wt% in chloroform (25 c) | |
COC1=CC=CC(OC)=C1C(O)=O | |
182.18 | |
182.18 | |
Crystalline Powder |
98.5 | |
187°C to 190°C | |
Authentic | |
C9H10O4 | |
MFCD00002437 | |
benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
MBIZFBDREVRUHY-UHFFFAOYSA-N | |
2,6-dimethoxybenzoic acid | |
15109 | |
99% | |
2, 6-Dimethoxybenzoic acid, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 215-985-4
missing translation for 'rtecsNumber' : DG8598725
missing translation for 'tsca' : TSCA
- 2,6-Dimethoxybenzoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only