Learn More
2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals
£28.84 - £88.14
Chemical Identifiers
CAS | 91-10-1 |
---|---|
Molecular Formula | C8H10O3 |
Molecular Weight (g/mol) | 154.17 |
MDL Number | MFCD00064434 |
InChI Key | KLIDCXVFHGNTTM-UHFFFAOYSA-N |
Synonym | syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol |
PubChem CID | 7041 |
ChEBI | CHEBI:955 |
IUPAC Name | 2,6-dimethoxyphenol |
SMILES | COC1=CC=CC(OC)=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11411307
|
Thermo Scientific Alfa Aesar
B24253.14 |
25 g |
£28.84
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11421307
|
Thermo Scientific Alfa Aesar
B24253.22 |
100 g |
£88.14
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
91-10-1 | |
154.17 | |
KLIDCXVFHGNTTM-UHFFFAOYSA-N | |
7041 | |
2,6-dimethoxyphenol |
C8H10O3 | |
MFCD00064434 | |
syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol | |
CHEBI:955 | |
COC1=CC=CC(OC)=C1O |
Specifications
91-10-1 | |
261°C | |
C8H10O3 | |
1526871 | |
Soluble in water. | |
COC1=CC=CC(OC)=C1O | |
154.17 | |
CHEBI:955 | |
99% |
50°C to 57°C | |
140°C (284°F) | |
MFCD00064434 | |
syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol | |
KLIDCXVFHGNTTM-UHFFFAOYSA-N | |
2,6-dimethoxyphenol | |
7041 | |
154.16 | |
2,6-Dimethoxyphenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 202-041-1
missing translation for 'rtecsNumber' : SL0900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only