missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethyl-4-pyrone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 132°C to 136°C |
Boiling Point | 248°C to 250°C |
Quantity | 250 g |
Beilstein | 107418 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 124.14 |
Percent Purity | 99% |
Chemical Name or Material | 2,6-Dimethyl-4-pyrone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'